EMAIL THIS PAGE TO A FRIEND
Acta obstetricia et gynecologica Scandinavica
Combined bladder and uterine rupture following prostaglandin-induced midtrimester abortion: an unreported event.
PMID 12956853
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000475
Carboprost trometamol, European Pharmacopoeia (EP) Reference Standard
C25H47NO8