EMAIL THIS PAGE TO A FRIEND
Guang pu xue yu guang pu fen xi = Guang pu
[Synthesis and characterization of complex of NdL3.2H2O].
PMID 12958944
Abstract
The binary solid complexes using rare earth ion Nd3+ with the ligand L (L = O-chorophenoxyacetic acid and 3-methoxybenzoic acid) has been synthesized in this work. The chemical formula of the complex was determined to be NdL3.2H2O by elemental analysis. The composition structure and properties of this type of complex were tentatively investigated by the analysis of molar conductivity, TG-DTA, IR and UV.