EMAIL THIS PAGE TO A FRIEND
Journal de toxicologie clinique et experimentale
[Chemical burns of the upper digestive tract and cytolytic hepatitis following ingestion of a mixture of methyl ethyl ketone peroxide, diacetone alcohol and diisobutyl phthalate].
PMID 1295970