EMAIL THIS PAGE TO A FRIEND
Guang pu xue yu guang pu fen xi = Guang pu
[Infrared spectroscopic investigation on complexes formed from cupric fluoborate and cupric butyrate with Bis (diphenylphosphino) alkane ligands].
PMID 12961872
Abstract
The Bis (diphenylphosphino) alkane series ligands and its series of complexes formed from Cu(BF4)2 and Cu(C3H7COO)2 respectively were studied by infrared spectral analysis. This paper discusses the spectral classification and the change regularity of related spectra before and after the Cu(I) complexes were formed. The possible structure model of the formed complexes was discussed, based on the element analysis, powder X-ray diffraction analysis, and thermal gravity analysis.
Related Materials