EMAIL THIS PAGE TO A FRIEND
Medical mycology
A case of kerion celsi due to Arthroderma benhamiae identified by DNA sequences of nuclear ribosomal internal transcribed spacer 1 regions.
PMID 12964717
Abstract
We describe a case of a 4-year-old boy with a 1-month history of a purulent lesion on his scalp. His hair samples revealed fungal organisms and Trichophyton mentagrophytes was cultured from the sample. We analysed the DNA sequences of the nuclear ribosomal internal transcribed spacer 1 (ITS1) region of the isolated fungus. These sequences were in accordance with T. mentagrophytes animal 4 type. In mating experiments, our strain only responded to the Arthroderma benhamiae Americano-European race (+) mating type tester. We speculate that the patient was infected from contact with his pet guinea pig. This is the first case of a clinical isolate of A. benhamiae being identified by DNA sequences of nuclear ribosomal ITS1 regions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32991
Lanoconazole, VETRANAL, analytical standard
C14H10ClN3S2