EMAIL THIS PAGE TO A FRIEND
Clinical chemistry and laboratory medicine
Reference values of serum total sialic acid for healthy adult subjects and comparison of two different calibrators for quantitation of total sialic acid in high performance anion-exchange chromatography with pulsed amperometric detection.
PMID 12964820
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8681
3′-Sialyllactose, from bovine milk or colostrum, ≥97% (HPLC)
C23H39NO19