EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Spectrofluorimetric determination of sulphonamides in pharmaceutical compounds and foods.
PMID 1298386
Abstract
Spectrofluorimetry and room temperature photochemically-induced fluorescence (RTPF) have been applied to the determination of sulphacetamide (SAC), sulphaguanidine (SG) and sulphamethazine (SMT) in milk and pharmaceutical formulations. The methods are suitable for determining 0.02-0.10 micrograms ml of SAC, 0.10-0.50 micrograms ml of SG, and 0.40-1.00 micrograms of SMT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S8751
Sulfaguanidine
C7H10N4O2S
32402
Sulfaguanidine, VETRANAL, analytical standard
C7H10N4O2S