EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Evaluation of mutagenic activity of some chemical combinations].
PMID 1298703
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
476315
4,4′-Diaminobenzanilide, 98%
C13H13N3O