EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Histochemical demonstration of aryl sulfatase.
PMID 13015105
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
190608
6-Benzoyl-2-naphthol, ≥98%
C17H12O2