EMAIL THIS PAGE TO A FRIEND
Human mutation
Human hepatic lipase mutations and polymorphisms.
PMID 1301939
Abstract
Human hepatic lipase (HL) is a 477 residue glycoprotein that hydrolyzes triglycerides from plasma lipoproteins. Familial HL deficiency is a rare recessive disorder that is characterized by premature atherosclerosis and abnormal circulating lipoproteins. While studying the HL gene from the world's index family with HL deficiency, we identified four coding sequence variants of HL, one in each of exons 4, 5, 6, and 8. In this report we present the genetic basis for two new HL gene variants, one in each of exons 3 and 5. All six HL DNA variants are single base pair changes. Two variants (at codons 133 and 202) are diallelic DNA polymorphisms that are silent at the amino acid level. One variant (V73M) is an allele that defines an uncommon HL isoprotein. One variant (N193S) has two alleles of approximately equal frequency in the population that specify two common HL isoproteins. Two variants (S267F and T383M) are rare mutations found to date only in HL deficient subjects and their relatives. Of the six HL variants described to date, only S267F and T383M are associated with hyperlipidemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
62287 Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
L4777
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
62288 Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
62301 Lipase from Aspergillus niger, powder (fine), ~200 U/g
L4277 Lipase from Aspergillus oryzae, ≥20,000 U/g
62285 Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
75577 Lipase from Burkholderia sp., lyophilized powder, slightly beige, ~12 U/mg
02569 Lipase from Candida antarctica, lyophilized, powder, beige, ~0.3 U/mg
62302 Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
L1754 Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
62316 Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
L8525 Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
L3170
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
L8906 Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
L9031 Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
62298 Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
L0382 Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
62309 Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
28602 Lipase from Pseudomonas fluorescens, powder, slightly beige, ≥160 U/mg
L9518
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
62310 Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
L3001 Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg protein
62306 Lipase from wheat germ, ~0.1 U/mg
73940 Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg