EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Information on basics of MAC for m-toluate iodide in the air of work area].
PMID 1305551
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
130737
3-Iodotoluene, 99%
C7H7I