EMAIL THIS PAGE TO A FRIEND
Folia biologica
Cell proliferation in the germinal layer of the lateral brain ventricles in normal adult mice and under some experimental conditions.
PMID 1306507
Abstract
Histological and autoradiographic studies revealed mitotic activity in the subependymal germinal layer in the brains of adult mice. The number of mitoses observed was higher in animals subjected to administration of haematoporphyrin and selenourea.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
230499
Selenourea, 98%
CH4N2Se