EMAIL THIS PAGE TO A FRIEND
Schriftenreihe des Vereins fur Wasser-, Boden- und Lufthygiene
[The algae toximeter for continuous water monitoring].
PMID 1307800
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36174
Monuron, PESTANAL®, analytical standard
C9H11ClN2O