EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The oxidative pathway of carbohydrate metabolism in Escherichia coli. 1. The isolation and properties of glucose 6-phosphate dehydrogenase and 6-phosphogluconate dehydrogenase.
PMID 13093611