EMAIL THIS PAGE TO A FRIEND
Food additives and contaminants
Estimation of the uncertainty in a multiresidue method for the determination of pesticide residues in fruit and vegetables.
PMID 13129793
Abstract
The estimation of uncertainty of an analytical result has become important in analytical chemistry. It is especially difficult to determine uncertainties for multiresidue methods, e.g. for pesticides in fruit and vegetables, as the varieties of pesticide/commodity combinations are many. In the present study, recommendations from the International Organisation for Standardisation's (ISO) Guide to the Expression of Uncertainty and the EURACHEM/CITAC guide Quantifying Uncertainty in Analytical Measurements were followed to estimate the expanded uncertainties for 153 pesticides in fruit and vegetables. Data from in-house validation were used in the estimation of the uncertainty. No significant difference in the relative standard deviation for reproducibility (RSD(R)) were found between the different concentration levels at concentration levels exceeding 2.5 times the detection limit. Therefore, it was possible to pool the RSD(R) within a single matrix. However, a difference in RSD(R) between matrices was seen, thus the poorest RSD(R) of the investigated matrices was chosen for the uncertainty estimation. The expanded uncertainties ranged from 7 to 78% with an average of 32% and median of 32%. Furthermore, only RSD(R) contributed to the uncertainty estimation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45477
Ethion, PESTANAL®, analytical standard
C9H22O4P2S4