EMAIL THIS PAGE TO A FRIEND
The Journal of physiology
An analysis of the central and peripheral components of respiratory failure produced by anticholinesterase poisoning in the rabbit.
PMID 13212728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32434
TEPP, PESTANAL®, analytical standard
C8H20O7P2