EMAIL THIS PAGE TO A FRIEND
Blood
The application of the Prussian blue stain to previously stained films of blood and bone marrow.
PMID 13230168
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
03899
Prussian blue soluble, for microscopy
C6Fe2KN6 · xH2O