EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Therapeutic factors in survival after lethal cholinesterase inhibition by phosphorus insecticides.
PMID 13244813
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32434
TEPP, PESTANAL®, analytical standard
C8H20O7P2