EMAIL THIS PAGE TO A FRIEND
Rassegna internazionale di clinica e terapia
[Intradermal test with mercuric oxycyanide in determination of the physical resistance of surgical patients].
PMID 13245994
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208140
Mercury(II) cyanide, 99%
C2HgN2