EMAIL THIS PAGE TO A FRIEND
British medical journal
Need for repeated microscopical tests in gonorrhoea.
PMID 13269913
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208140
Mercury(II) cyanide, 99%
C2HgN2