EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
The histochemical demonstration of acid phosphatase by a post-incubation coupling technique.
PMID 13271750
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
190608
6-Benzoyl-2-naphthol, ≥98%
C17H12O2