EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Differential effects of methylmercury, thiols, and vitamins on galactosidases of nervous and non-nervous tissues.
PMID 1327315
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A16602
DL-N-Acetylhomocysteine thiolactone, 98%
C6H9NO2S