EMAIL THIS PAGE TO A FRIEND
The Journal of membrane biology
Sulfhydryl alkylating agents induce calcium current in skeletal muscle fibers of a crustacean (Atya lanipes).
PMID 1331464
Abstract
Voltage-clamp experiments using the three-microelectrode voltage clamp technique were performed on ventroabdominal flexor muscles of the crustacean Atya lanipes. Potassium and chloride currents were found to underlie the normal, passive response of the muscle. Blocking potassium currents with tetraethylammonium and replacing chloride ions with methanesulfonate did not unmask an inward current. By treating the muscle with the sulfhydryl-alkylating agent 4-cyclopentene-1,3-dione an inward current was detected. The current induced by the agent is carried by Ca2+, since it is abolished in Ca(2+)-free solutions. The induced Ca2+ current is detected at about -40 mV and reaches a mean maximum value of -78 microA/cm2 at ca. -10 mV. At this potential the time to peak is close to 15 msec. The induced Ca2+ current inactivated with 1-sec prepulses which did not elicit detectable Ca2+ current; the fitted hx curve had a midpoint of -38 mV and a steepness of 5.0 mV. Measurements of isometric tension were performed in small bundles of fibers, and the effects of the sulfhydryl-alkylating agents 4-cyclopentene-1,3-dione and N-ethylmaleimide were investigated. Tetanic tension was enhanced in a strictly Ca(2+)-dependent manner by 4-cyclopentene-1,3-dione. The amplitude of K+ contractures increased after treatment with N-ethylmaleimide. It is concluded that Ca2+ channels are made functional by the sulfhydryl-specific reagents and that the increase in tension is probably mediated by an increase in Ca2+ influx through the chemically induced Ca2+ channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161683
4-Cyclopentene-1,3-dione, 95%
C5H4O2