EMAIL THIS PAGE TO A FRIEND
Vestnik oftalmologii
[Use of a new Soviet miotic, the tetraethylthiopyrophosphate (A2) in glaucoma].
PMID 13325236
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32434
TEPP, PESTANAL®, analytical standard
C8H20O7P2