EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Syntheses of specifically deoxygenated methyl alpha-isomaltotriosides.
PMID 1332823
Abstract
Specifically deoxygenated methyl alpha-isomaltotriosides were synthesized by the silver perchlorate-mediated condensation of a protected alpha-isomaltosyl chloride with suitably blocked derivatives of methyl alpha-D-glucopyranoside deoxygenated respectively at positions 2, 3, and 4, followed by deprotection.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
674583
Silver perchlorate, anhydrous, 97%
AgClO4