EMAIL THIS PAGE TO A FRIEND
Protein expression and purification
Isolation of amine oxidase from bovine plasma by a two-step procedure.
PMID 1333859
Abstract
A novel method for isolation of amine oxidase from bovine plasma is reported; it involves a two-step procedure, namely ammonium sulfate fractionation and affinity chromatography with elution by aniline, which is a competitive inhibitor of the enzyme. A homogeneous enzyme, characterized by a specific activity of 0.44 U/mg, was obtained with a yield higher than 50%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6017
ω-Aminohexyl–Agarose, saline suspension