EMAIL THIS PAGE TO A FRIEND
Progress in lipid research
Gamma linolenic acid--occurrence and physical and chemical properties.
PMID 1334265
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2378
γ-Linolenic acid, ≥99%, liquid
C18H30O2
62174
γ-Linolenic acid, analytical standard
C18H30O2