EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
The mechanism of action of chlorhexidine.
PMID 1335944
Abstract
Chlorhexidine did not inhibit ATPase in intact cells of Escherichia coli K12 W1317i-, even at bactericidal concentrations, and ATP hydrolysis was greatest at the highest concentration (40 mg/l), even though no net uptake of substrate occurred. Like dinitrophenol and tribrominated salicylanilide, polymyxin and chlorhexidine collapsed the membrane potential at inhibitory concentrations. Membrane disruption, and not ATPase inactivation, is considered the lethal event in chlorhexidine action.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8146
Methyl-β-D-thiogalactoside
C7H14O5S