EMAIL THIS PAGE TO A FRIEND
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.)
Propagation of strain L (Earle) cells in agitated fluid suspension cultures.
PMID 13370534
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
L-M, 87032401