EMAIL THIS PAGE TO A FRIEND
Journal of biochemical and biophysical methods
Photochrome-labeling method in study of dynamics of biological systems.
PMID 1337353
Abstract
The theoretical considerations and experimental evidences discussed in this paper indicate that quantitative study of photochromic processes in labeled objects open up new possibilities for investigating microviscosity and conformation transitions in biological systems. The proposed method of photochrome labeling features higher sensitivity and simplicity, and uses labels which are more stable under physiological conditions compared with traditional spin labels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
253367
4-(2-Iodoacetamido)-TEMPO, free radical
C11H20IN2O2