EMAIL THIS PAGE TO A FRIEND
Cancer research
Further studies on the carcinogenicity of dyes related to 4-dimethylaminoazobenzene; the requirement for an unsubstituted 2-position.
PMID 13437302
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
263532
3,5-Difluoroaniline, 98%
C6H5F2N