EMAIL THIS PAGE TO A FRIEND
The American journal of pathology
Histologic and physiologic characteristics of hormone-secreting transplantable adrenal tumors in mice and rats.
PMID 13444454
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
Y1, 85062807