EMAIL THIS PAGE TO A FRIEND
Bulletins et memoires de la Societe medicale des hopitaux de Paris
[Treatment of primary-secondary syphilis in adults by 15,000,000 units of penicillin in a single series preceded by mercury cyanide; statement of statistics on re-infection up to maximum of 9 1/2 years].
PMID 13446703
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208140
Mercury(II) cyanide, 99%
C2HgN2