EMAIL THIS PAGE TO A FRIEND
Natural toxins
Pyrrolizidine alkaloids from Melampyrum pratense.
PMID 1344898
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73913
Seneciphylline, analytical standard
C18H23NO5