EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Robust dismissal of a claim on chemical sensitisation.
PMID 1346295
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W203203
Allyl hexanoate, ≥98%, FCC, FG
C9H16O2
W203211
Allyl hexanoate, natural, ≥98%, FCC, FG
C9H16O2