EMAIL THIS PAGE TO A FRIEND
Metabolism: clinical and experimental
Hypoglycemic effects of a beta-agonist, Ro 16-8714, in streptozotocin-diabetic rats: decreased hepatic glucose production and increased glucose utilization in oxidative muscles.
PMID 1346545
Abstract
Streptozotocin (STZ)-induced diabetic rats are glycosuric, hyperglycemic, hyperketonemic, overproduce glucose, and have a decreased glucose utilization in oxidative muscles. Treatment with a beta-agonist, Ro 16-8714, decreases the glycosuria, hyperglycemia, hyperketonemia, and hepatic glucose production. Tissue glucose utilization was unchanged, except in oxidative muscles, where it was increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A72405
2-Amino-1-phenylethanol, 98%
C8H11NO