EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
3-Deaza-adenosine and inhibition of HIV.
PMID 1346708
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8296
3-Deazaadenosine
C11H14N4O4