EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Return to Pakistan of pipenzolate plus phenobarbitone.
PMID 1346854
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6085
Pipenzolate bromide, BioXtra
C22H28BrNO3
P8545
Pipenzolate bromide, powder
C22H28BrNO3