EMAIL THIS PAGE TO A FRIEND
British journal of cancer
The sex chromatin in human malignant tissues.
PMID 13499787
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0290 Hartman’s Fixative, histological tissue fixative