EMAIL THIS PAGE TO A FRIEND
Bulletins et memoires de la Societe medicale des hopitaux de Paris
[Treatment of primary & secondary syphilis by penicillin & mercuric cyanide].
PMID 13511154
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208140
Mercury(II) cyanide, 99%
C2HgN2