EMAIL THIS PAGE TO A FRIEND
The Journal of Comparative Neurology
Neurotensin terminals form synapses primarily with neurons lacking detectable tyrosine hydroxylase immunoreactivity in the rat substantia nigra and ventral tegmental area.
PMID 1351897
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB5496 Anti-Neurotensin Antibody