EMAIL THIS PAGE TO A FRIEND
Hoppe-Seyler's Zeitschrift fur physiologische Chemie
[Further studies on the metabolic behavior of some aromatic and hydroaromatic mono- and dicarboxylic acids].
PMID 13548876
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T38091
m-Tolylacetic acid, 99%
C9H10O2