EMAIL THIS PAGE TO A FRIEND
Klinische Medizin; osterreichische Zeitschrift fur wissenschaftliche und praktische Medizin
[External therapy of chronic dermatoses].
PMID 13550751
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
131768
Salicylanilide, 98%
C13H11NO2