EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Synthesis of 1233A analogs and their inhibitory activity against hydroxymethylglutaryl coenzyme A synthase.
PMID 1355474
Abstract
Simple and efficient syntheses of 1233A analogs were developed and the inhibitory activity of the analogs against hydroxymethylglutaryl coenzyme A (HMG-CoA) synthase was determined. Study of the structure-activity relationships revealed that not only the geometry in beta-lactone moiety but also the length of the carbon side chain is important for inhibitory activity against HMG-CoA synthase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0301 Hymeglusin, ≥98% (HPLC), from Fusarium sp.
C18H28O5