EMAIL THIS PAGE TO A FRIEND
Acta tropica
Antigenic mimicry between piperazine derivatives and Wuchereria bancrofti microfilariae.
PMID 1356307
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
335924
1-Piperidinepropionic acid, 96%
C8H15NO2