EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Enzymatic synthesis of deoxyribonucleic acid. I. Preparation of substrates and partial purification of an enzyme from Escherichia coli.
PMID 13563462