EMAIL THIS PAGE TO A FRIEND
Biochemische Zeitschrift
[Phosphomannose isomerase. I. Activity measurement and dependence of enzyme action on sulfhydryl groups and metals in some animal tissues].
PMID 13596383