EMAIL THIS PAGE TO A FRIEND
Neuroscience letters
Sulmazole effects on PGE2 and D-Ala2-Met-enkephalinamide modulation of cyclic AMP synthesis and neurotransmitter release in a sympathetic ganglion.
PMID 1361043
Abstract
In the guinea-pig superior cervical ganglion, the Gi blocking agent sulmazole enhanced the basal and prostaglandin E2-induced stimulation of cyclic AMP synthesis but had no effect on the prostaglandin-dependent inhibition of acetylcholine release. On the contrary sulmazole counteracted the inhibitory effect of D-Ala2-Met-enkephalinamide both on cyclic AMP formation and acetylcholine release. Moreover sulmazole eliminated the supra-additive effect of the combination of prostaglandin + opiate on cyclic AMP synthesis. The presence of a Gi-coupled opiate receptor at the pre-and postsynaptic levels is suggested.