EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Bacterial oxidation of vitamin B6. I. Isopyridoxal and 5-pyridoxic acid.
PMID 13610871
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74006
Isopyridoxal hydrochloride, ≥98.5% (HPLC)
C8H9NO3· HCl