EMAIL THIS PAGE TO A FRIEND
Human & experimental toxicology
Difenacoum poisoning as a cause of haematuria.
PMID 1361149
Abstract
A man presented with frank haematuria and a grossly prolonged prothrombin time. He was later found to have taken an overdose of difenacoum--a 'superwarfarin' rodenticide. The diagnosis was confirmed by a serum concentration of difenacoum of 0.6 micrograms ml-1. Overdosage with superwarfarins is discussed and the need for prolonged treatment with vitamin K1 highlighted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32677
Difenacoum, PESTANAL®, analytical standard
C31H24O3